2,5-Dimethoxytoluol 95%
Lieferant: Apollo Scientific
Synonyme:
1,4-Dimethoxy-2-methylbenzol
OR925248-25G
APOSOR925248-25GEA
95.7
EUR
APOSOR925248-25G
2,5-Dimethoxytoluol 95%
2,5-Dimethoxytoluol
Formel:
C₉H₁₂O₂ Molecular Weight: 152,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041901 CAS-Nummer: 24599-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...