1-Naphthoesäure 98%
Lieferant: Apollo Scientific
Synonyme:
Naphthalin-1-carbonsäure, α-Naphthalincarbonsäure, 1-Naphthalincarbonsäure, α-Naphthoesäure
OR925255-500G
APOSOR925255-500GEA
121
EUR
APOSOR925255-500G
1-Naphthoesäure 98%
1-Naphthoesäure
Formel:
C₁₁H₈O₂ Molecular Weight: 172,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004007 CAS-Nummer: 86-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...