Diethylammoniumchlorid 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: Diethylammonium chloride

OR925271-5KG OR925271-1KG OR925271-250G
APOSOR925271-5KGEA 546 EUR
APOSOR925271-5KG APOSOR925271-1KG APOSOR925271-250G
Diethylammoniumchlorid 99%
Diethylammoniumchlorid
Formel: C₄H₁₂ClN
Molecular Weight: 109,6 g/mol
Siedepunkt: 320…330 °C/760mmHg
Schmelzpunkt: 227…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012499
CAS-Nummer: 660-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR