4-Phenoxybenzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Phenoxyphenyl)methanol

OR9253-1G OR9253-5G
APOSOR9253-1GEA 158 EUR
APOSOR9253-1G APOSOR9253-5G
4-Phenoxybenzyl alcohol 97%
4-Phenoxybenzyl alcohol
Formel: C₁₃H₁₂O₂
Molecular Weight: 200,237 g/mol
Schmelzpunkt: 55…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01463970
CAS-Nummer: 2215-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR