(2S,3S)-2-Chlor-3-methylpentansäure 95%
Lieferant: Apollo Scientific
Synonyme:
(2S,3S)-2-Chloro-3-methylpentanoic Acid
OR925320-1G
OR925320-5G
APOSOR925320-1GEA
335
EUR
APOSOR925320-1G
APOSOR925320-5G
(2S,3S)-2-Chlor-3-methylpentansäure 95%
(2S,3S)-2-Chloro-3-methylvaleric acid
Formel:
C₆H₁₁ClO₂ Molecular Weight: 150,60 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191403 CAS-Nummer: 32653-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...