1-(Methoxymethoxy)naphthalin 95%
Lieferant: Apollo Scientific
OR925344-10G
OR925344-1G
OR925344-5G
APOSOR925344-10GEA
705
EUR
APOSOR925344-10G
APOSOR925344-1G
APOSOR925344-5G
1-(Methoxymethoxy)naphthalin 95%
1-(Methoxymethoxy)naphthalene
Formel:
C₁₂H₁₂O₂ Molecular Weight: 188,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797119 CAS-Nummer: 7382-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...