(3-Methoxybenzyl)methylamine 95%

Lieferant: Apollo Scientific

Synonyme: 3-Methoxy-N-methylbenzylamine, 1-(3-Methoxyphenyl)-N-methylmethanamine

OR925353-5G
APOSOR925353-5GEA 71.2 EUR
APOSOR925353-5G
(3-Methoxybenzyl)methylamine 95%
(3-Methoxybenzyl)methylamine
Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04618244
CAS-Nummer: 41789-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR