5-Methylnicotinamid 95%

Lieferant: Apollo Scientific

OR925361-1G OR925361-5G
APOSOR925361-1GEA 183 EUR
APOSOR925361-1G APOSOR925361-5G
5-Methylnicotinamid 95%
5-Methylnicotinamid
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664041
CAS-Nummer: 70-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR