1-Naphthyl(oxo)acetonitril 95%
Lieferant: Apollo Scientific
Synonyme:
1-Naphthalenecarbonyl Cyanide
OR925363-10G
OR925363-1G
OR925363-5G
APOSOR925363-10GEA
886
EUR
APOSOR925363-10G
APOSOR925363-1G
APOSOR925363-5G
1-Naphthyl(oxo)acetonitril 95%
1-Naphthoyl cyanide
Formel:
C₁₂H₇NO Molecular Weight: 181,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060239 CAS-Nummer: 14271-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...