Isopentylnitrat 95%

Lieferant: Apollo Scientific

Synonyme: Isoamylnitrat

OR925387-25G OR925387-5G
APOSOR925387-25GEA 299 EUR
APOSOR925387-25G APOSOR925387-5G
Isopentylnitrat 95%
Isopentylnitrat
Formel: C₅H₁₁NO₃
Molecular Weight: 133,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007042
CAS-Nummer: 543-87-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR