1-(3-Chlor-2-pyridinyl)piperazin 98%

Lieferant: Apollo Scientific

OR925578-10G OR925578-1G OR925578-5G
APOSOR925578-10GEA 928 EUR
APOSOR925578-10G APOSOR925578-1G APOSOR925578-5G
1-(3-Chlor-2-pyridinyl)piperazin 98%
1-(3-Chlor-2-pyridinyl)piperazin
Formel: C₉H₁₂ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08061034
CAS-Nummer: 87394-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR