2-(Thiazol-4-yl)acetic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Thiazolyl)acetic Acid

OR925603-1G OR925603-5G
APOSOR925603-1GEA 106 EUR
APOSOR925603-1G APOSOR925603-5G
2-(Thiazol-4-yl)acetic acid 97%
2-(Thiazol-4-yl)acetic acid
Formel: C₅H₅NO₂S
Molecular Weight: 143,16 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09991935
CAS-Nummer: 7504-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR