Furfuryl-methyldisulfid 97%

Lieferant: Apollo Scientific

Synonyme: 2-((Methyldisulphanyl)methyl)furan, Methyl furfuryl disulphide

OR925998-100G
APOSOR925998-100GEA 60.9 EUR
APOSOR925998-100G
Furfuryl-methyldisulfid 97%
Furfuryl-methyldisulfid
Formel: C₆H₈OS₂
Molecular Weight: 160,2571 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012326
CAS-Nummer: 57500-00-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR