3,5-Dibromo-2-chloropyrazine ≥98%

Lieferant: Apollo Scientific

OR926025-5G
APOSOR926025-5GEA 94.4 EUR
APOSOR926025-5G
3,5-Dibromo-2-chloropyrazine ≥98%
3,5-Dibromo-2-chloropyrazine
Formel: C4HBr2ClN2
Molecular Weight: 272,3300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13193367
CAS-Nummer: 1082843-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR