3,5-Dimethylpyrazin-2-ol 95%

Lieferant: Apollo Scientific

OR926083-10G OR926083-1G OR926083-5G
APOSOR926083-10GEA 503 EUR
APOSOR926083-10G APOSOR926083-1G APOSOR926083-5G
3,5-Dimethylpyrazin-2-ol 95%
3,5-Dimethylpyrazin-2-ol
Formel: C6H8N2O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 60187-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR