3,5-Dimethyl-1H-indole-2-carbaldehyde 95%

Lieferant: Apollo Scientific

OR926109-1G OR926109-5G
APOSOR926109-1GEA 219 EUR
APOSOR926109-1G APOSOR926109-5G
3,5-Dimethyl-1H-indole-2-carbaldehyde 95%
3,5-Dimethyl-1H-indole-2-carbaldehyde
Formel: C₁₁H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD07643177
CAS-Nummer: 1463-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR