2-Ethyl-3,5-dimethylpyridin 95%

Lieferant: Apollo Scientific

OR926270-1G OR926270-5G
APOSOR926270-1GEA 313 EUR
APOSOR926270-1G APOSOR926270-5G
2-Ethyl-3,5-dimethylpyridin 95%
2-Ethyl-3,5-dimethylpyridin
Formel: C₉H₁₃N
Molecular Weight: 135,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051049
CAS-Nummer: 1123-96-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR