4-Methoxy-2,5-dimethyl-3(2H)-furanon 95%

Lieferant: Apollo Scientific

Synonyme: 4-Methoxy-2,5-dimethylfuran-3(2H)-one, 4-Methoxy-2,5-dimethyl-3(2H)-furanone

OR926305-25G
APOSOR926305-25GEA 71.2 EUR
APOSOR926305-25G
4-Methoxy-2,5-dimethyl-3(2H)-furanon 95%
2,5-Dimethyl-4-methoxy-3(2H)-furanone
Formel: C₇H₁₀O₃
Molecular Weight: 142,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00209504
CAS-Nummer: 4077-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR