5-Methylthiazole-4-carboxylic acid ethyl ester 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 5-methylthiazole-4-carboxylate
OR926418-1G
OR926418-250MG
APOSOR926418-1GEA
78.3
EUR
APOSOR926418-1G
APOSOR926418-250MG
5-Methylthiazole-4-carboxylic acid ethyl ester 97%
5-Methylthiazole-4-carboxylic acid ethyl ester
Formel:
C₇H₉NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD18427704 CAS-Nummer: 61323-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...