H-D-Abu-OtBu·HCl 97%

Lieferant: Apollo Scientific

Synonyme: H-D-Abu-OtBu·HCl

OR926472-1G OR926472-5G
APOSOR926472-1GEA 53.8 EUR
APOSOR926472-1G APOSOR926472-5G
H-D-Abu-OtBu·HCl 97%
H-D-Abu-OtBu·HCl
Formel: C₈H₁₈ClNO₂
Molecular Weight: 195,687 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08275809
CAS-Nummer: 313994-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR