2-Chlor-4-nitrotoluol 98%

Lieferant: Apollo Scientific

OR926495-100G OR926495-500G
APOSOR926495-100GEA 55.1 EUR
APOSOR926495-100G APOSOR926495-500G
2-Chlor-4-nitrotoluol 98%
2-Chlor-4-nitrotoluol
Formel: C₇H₆ClNO₂
Molecular Weight: 171,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007210
CAS-Nummer: 121-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR