4-Amino-2-methoxybenzonitril 98%
Lieferant: Apollo Scientific
OR926533-1G
OR926533-5G
APOSOR926533-1GEA
88.6
EUR
APOSOR926533-1G
APOSOR926533-5G
4-Amino-2-methoxybenzonitril 98%
4-Amino-2-methoxybenzonitril
Formel:
C₈H₈N₂O Molecular Weight: 148,1619 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027303 CAS-Nummer: 7251-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...