Ethylcyclohexylacetat 97%

Lieferant: Apollo Scientific

Synonyme: Cyclohexylacetic acid ethyl ester, Ethyl Cyclohexaneacetate, Cyclohexaneacetic acid ethyl ester

OR926544-100G OR926544-25G OR926544-5G
APOSOR926544-100GEA 647 EUR
APOSOR926544-100G APOSOR926544-25G APOSOR926544-5G
Ethylcyclohexylacetat 97%
Ethylcyclohexylacetat
Formel: C₁₀H₁₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00001517
CAS-Nummer: 5452-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR