3-Methyl-1,3-thiazolane-2,4-dione 95%

Lieferant: Apollo Scientific

Synonyme: 3-Methylthiazolidine-2,4-dione

OR926563-1G OR926563-250MG OR926563-5G
APOSOR926563-1GEA 72.5 EUR
APOSOR926563-1G APOSOR926563-250MG APOSOR926563-5G
3-Methyl-1,3-thiazolane-2,4-dione 95%
3-Methyl-1,3-thiazolane-2,4-dione
Formel: C₄H₅NO₂S
Molecular Weight: 131,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02194457
CAS-Nummer: 16312-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR