2,4-Dimethylpyrimidin-5-ol

Lieferant: Apollo Scientific
Achtung

OR926618-100MG OR926618-1G OR926618-250MG
APOSOR926618-100MGEA 52.2 EUR
APOSOR926618-100MG APOSOR926618-1G APOSOR926618-250MG
2,4-Dimethylpyrimidin-5-ol
2,4-Dimethylpyrimidin-5-ol
Formel: C₆H₈N₂O
Molecular Weight: 124,1400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18072447
CAS-Nummer: 412003-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR