6-Methyl-4-chromanone 95%
Lieferant: Apollo Scientific
Synonyme:
6-Methylchroman-4-one
OR926669-5G
APOSOR926669-5GEA
119
EUR
APOSOR926669-5G
6-Methyl-4-chromanone 95%
6-Methyl-4-chromanone
Formel:
C₁₀H₁₀O₂ Molecular Weight: 162,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024071 CAS-Nummer: 39513-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...