2-m-Tolylpyrimidine-5-carbaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 2-(3-Methylphenyl)pyrimidine-5-carbaldehyde

OR926685-1G OR926685-5G
APOSOR926685-1GEA 139 EUR
APOSOR926685-1G APOSOR926685-5G
2-m-Tolylpyrimidine-5-carbaldehyde 95%
2-m-Tolylpyrimidine-5-carbaldehyde
Formel: C₁₂H₁₀N₂O
Molecular Weight: 198,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059844
CAS-Nummer: 915921-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR