(2E)-3-(1,3,5-Trimethyl-1H-pyrazol-4-yl)acrylic acid 95%
Lieferant: Apollo Scientific
OR926712-1G
OR926712-250MG
APOSOR926712-1GEA
538
EUR
APOSOR926712-1G
APOSOR926712-250MG
(2E)-3-(1,3,5-Trimethyl-1H-pyrazol-4-yl)acrylic acid 95%
(2E)-3-(1,3,5-Trimethyl-1H-pyrazol-4-yl)acrylic acid
Formel:
C₉H₁₂N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00462197 CAS-Nummer: 21937-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...