2-Ethylquinazolin-4(3H)-one 95%
Lieferant: Apollo Scientific
OR926723-1G
APOSOR926723-1GEA
81.2
EUR
APOSOR926723-1G
2-Ethylquinazolin-4(3H)-one 95%
2-Ethylquinazolin-4(3H)-one
Formel:
C₁₀H₁₀N₂O Molecular Weight: 174,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00247024 CAS-Nummer: 3137-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...