2-Chlor-DL-phenylalanin 97%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-DL-phenylalanine, 2-Amino-3-(2-chlorophenyl)propanoic acid, 2-Chlorophenylalanine

OR926734-25G OR926734-5G
APOSOR926734-25GEA 133 EUR
APOSOR926734-25G APOSOR926734-5G
2-Chlor-DL-phenylalanin 97%
2-Chlor-DL-phenylalanin
Formel: C₉H₁₀ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00037774
CAS-Nummer: 14091-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR