2-(4-Methylpiperazin-1-ylmethyl)benzylamine 97%

Lieferant: Apollo Scientific

Synonyme: {2-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine

OR9268-1G OR9268-5G
APOSOR9268-1GEA 248 EUR
APOSOR9268-1G APOSOR9268-5G
2-(4-Methylpiperazin-1-ylmethyl)benzylamine 97%
2-(4-Methylpiperazin-1-ylmethyl)benzylamine
Formel: C₁₃H₂₁N₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 879896-50-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR