6-Chloro-quinolin-4-ylamine 95%

Lieferant: Apollo Scientific

Synonyme: 4-Amino-6-chloroquinoline

OR926895-1G OR926895-5G
APOSOR926895-1GEA 328 EUR
APOSOR926895-1G APOSOR926895-5G
6-Chloro-quinolin-4-ylamine 95%
6-Chloro-quinolin-4-ylamine
Formel: C₉H₇ClN₂
Molecular Weight: 178,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658304
CAS-Nummer: 20028-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR