4'-Chlor-2'-nitroacetophenon 98%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Chloro-2-nitrophenyl)ethanone

OR926954-5G
APOSOR926954-5GEA 260 EUR
APOSOR926954-5G
4'-Chlor-2'-nitroacetophenon 98%
4'-Chlor-2'-nitroacetophenon
Formel: C₈H₆ClNO₃
Molecular Weight: 199.5900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09031331
CAS-Nummer: 23082-51-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR