Ethyl-2-amino-5-methoxybenzoate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-amino-5-methoxybenzoate

OR926967-1G OR926967-5G
APOSOR926967-1GEA 63.8 EUR
APOSOR926967-1G APOSOR926967-5G
Ethyl-2-amino-5-methoxybenzoate 97%
Ethyl-2-amino-5-methoxybenzoate
Formel: C₁₀H₁₃NO₃
Molecular Weight: 195.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11111632
CAS-Nummer: 64018-98-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR