O-Methyl-L-tyrosine 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-2-Amino-3-(4-methoxyphenyl)propionic Acid, H-Tyr(Me)-OH, 4-Methoxy-L-phenylalanine
OR927182-25G
OR927182-5G
APOSOR927182-25GEA
589
EUR
APOSOR927182-25G
APOSOR927182-5G
O-Methyl-L-tyrosine 97%
O-Methyl-L-tyrosine
Formel:
C₁₀H₁₃NO₃ Molecular Weight: 195,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002604 CAS-Nummer: 6230-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...