3-Ethoxy-4-hydroxyphenylacetic acid 95%

Lieferant: Apollo Scientific

OR927302-1G OR927302-5G
APOSOR927302-1GEA 132 EUR
APOSOR927302-1G APOSOR927302-5G
3-Ethoxy-4-hydroxyphenylacetic acid 95%
3-Ethoxy-4-hydroxyphenylacetic acid
Formel: C₁₀H₁₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00016835
CAS-Nummer: 80018-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR