(2,5-Dimethoxyphenyl)essigsäure 95%

Lieferant: Apollo Scientific

OR927331-100G OR927331-25G OR927331-5G
APOSOR927331-100GEA 782 EUR
APOSOR927331-100G APOSOR927331-25G APOSOR927331-5G
(2,5-Dimethoxyphenyl)essigsäure 95%
(2,5-Dimethoxyphenyl)essigsäure
Formel: (C₆H₃(OCH₃)₂)CH₂COOH
Molecular Weight: 196,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004322
CAS-Nummer: 1758-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR