1,1,3,3-Tetramethyl-2-thioharnstoff 98%

Lieferant: Apollo Scientific

Synonyme: Tetramethylthioharnstoff, Tetramethyl-2-thioharnstoff, N,N,N',N'-Tetramethylthioharnstoff

OR927378-100G OR927378-25G OR927378-500G
APOSOR927378-100GEA 65.4 EUR
APOSOR927378-100G APOSOR927378-25G APOSOR927378-500G
1,1,3,3-Tetramethyl-2-thioharnstoff 98%
1,1,3,3-Tetramethyl-2-thioharnstoff
Formel: C₅H₁₂N₂S
Molecular Weight: 132,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008324
CAS-Nummer: 2782-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR