4-Methyl-1H-indole-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR927379-1G OR927379-250MG
APOSOR927379-1GEA 264 EUR
APOSOR927379-1G APOSOR927379-250MG
4-Methyl-1H-indole-3-carboxylic acid 95%
4-Methyl-1H-indole-3-carboxylic acid
Formel: C₁₀H₉NO₂
Molecular Weight: 175,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07776742
CAS-Nummer: 858515-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR