4-Azaspiro[2.4]heptan-5-one 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR927431-1G
OR927431-250MG
APOSOR927431-1GEA
510
EUR
APOSOR927431-1G
APOSOR927431-250MG
4-Azaspiro[2.4]heptan-5-one 95%
4-Azaspiro[2.4]heptan-5-one
Formel:
C₆H₉NO Molecular Weight: 111.14 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD15146411 CAS-Nummer: 308266-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...