5,6,7,8-Tetrahydro-2,4(1H,3H)-quinazolinedione 96%

Lieferant: Apollo Scientific

Synonyme: 1,2,3,4,5,6,7,8-Octahydroquinazoline-2,4-dione

OR927534-10G OR927534-1G OR927534-5G
APOSOR927534-10GEA 944 EUR
APOSOR927534-10G APOSOR927534-1G APOSOR927534-5G
5,6,7,8-Tetrahydro-2,4(1H,3H)-quinazolinedione 96%
5,6,7,8-Tetrahydro-2,4(1H,3H)-quinazolinedione
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13176330
CAS-Nummer: 35042-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR