Dimetridazol 98%

Lieferant: Apollo Scientific

Synonyme: 1,2-Dimethyl-5-nitroimidazole

OR927600-500G
APOSOR927600-500GEA 112 EUR
APOSOR927600-500G
Dimetridazol 98%
Dimetridazol
Formel: C₅H₇N₃O₂
Molecular Weight: 141,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047046
CAS-Nummer: 551-92-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR