2-Bromo-4-methylpentanoic acid 95%

Lieferant: Apollo Scientific

OR927629-1G
APOSOR927629-1GEA 68.3 EUR
APOSOR927629-1G
2-Bromo-4-methylpentanoic acid 95%
2-Bromo-4-methylpentanoic acid
Formel: C₆H₁₁BrO₂
Molecular Weight: 195,0543 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039606
CAS-Nummer: 42990-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR