tert-Butyl-2-hydroxy-2-methylpropylcarbamate 95%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 2-hydroxy-2-methylpropylcarbamate
OR927648-1G
APOSOR927648-1GEA
36.4
EUR
APOSOR927648-1G
tert-Butyl-2-hydroxy-2-methylpropylcarbamate 95%
tert-Butyl-2-hydroxy-2-methylpropylcarbamate
Formel:
C₉H₁₉NO₃ Molecular Weight: 189,2500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09608009 CAS-Nummer: 183059-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...