Nitrosobenzol 95%

Lieferant: Apollo Scientific

OR927665-100G OR927665-25G
APOSOR927665-100GEA 324 EUR
APOSOR927665-100G APOSOR927665-25G
Nitrosobenzol 95%
Nitrosobenzene
Formel: C₆H₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00002059
CAS-Nummer: 586-96-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR