N-Benzoyl-L-alanine 98%

Lieferant: Apollo Scientific

Synonyme: Bz-Ala-OH, (S)-2-Benzamidopropanoic acid

OR927719-25G OR927719-100G
APOSOR927719-25GEA 71.2 EUR
APOSOR927719-25G APOSOR927719-100G
N-Benzoyl-L-alanine 98%
N-Benzoyl-L-alanine
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193.202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063128
CAS-Nummer: 2198-64-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR