3-Methyl-2-thiophensulfonylchlorid 95%
Lieferant: Apollo Scientific
OR927734-1G
OR927734-5G
APOSOR927734-1GEA
183
EUR
APOSOR927734-1G
APOSOR927734-5G
3-Methyl-2-thiophensulfonylchlorid 95%
3-Methyl-2-thiophensulfonylchlorid
Formel:
C₅H₅ClO₂S₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864262 CAS-Nummer: 61714-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...