5-Ethylthiophene-3-carboxylic acid 95%
Lieferant: Apollo Scientific
OR927755-1G
OR927755-250MG
APOSOR927755-1GEA
623
EUR
APOSOR927755-1G
APOSOR927755-250MG
5-Ethylthiophene-3-carboxylic acid 95%
5-Ethylthiophene-3-carboxylic acid
Formel:
C₇H₈O₂S Molecular Weight: 156.2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419849 CAS-Nummer: 19156-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...