Methyl-o-anisat 95%

Lieferant: Apollo Scientific

Synonyme: Methyl-2-methoxybenzoat, 2-Methoxybenzoesäuremethylester, o-Anissäuremethylester, 2-Anissäuremethylester

OR927757-500G
APOSOR927757-500GEA 201 EUR
APOSOR927757-500G
Methyl-o-anisat 95%
Methyl-o-anisat
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008423
CAS-Nummer: 606-45-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR