6-Chloro-1H-[1,2,3]triazolo[4,5-b]pyridine 95%
Lieferant: Apollo Scientific
OR927905-1G
OR927905-250MG
APOSOR927905-1GEA
200
EUR
APOSOR927905-1G
APOSOR927905-250MG
6-Chloro-1H-[1,2,3]triazolo[4,5-b]pyridine 95%
6-Chloro-1H-[1,2,3]triazolo[4,5-b]pyridine
Formel:
C₅H₃ClN₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669468 CAS-Nummer: 408314-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...